Multiplex PCR 5X Master Mix , New England Biolabs
Supplier: New England Biolabs (NEB)
M0284S
101446-500EA 289 USD
101446-500
Multiplex PCR 5X Master Mix , New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The NEB Multiplex PCR 5X Master Mix is an easy-to-use solution featuring high quality recombinant Taq DNA Polymerase
The mix is optimized for high yield and robust performance in multiplex PCR reactions. The 5X formulation allows the user maximal input of primers, template DNAs and additional components.