Taq 5X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
M0285L
101444-748EA 171 USD
101444-748
Taq 5X Master Mix, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Taq 5X Master Mix is an optimized ready-to-use solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCl and stabilizers
It is best for routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA. It can amplify up to 4 kb from complex genomic DNA or up to 5 kb from lambda DNA.