Disodium malonate 2 M

Supplier: Rigaku Reagents

Synonyms: Sodium malonate dibasic, Malonic acid disodium salt

1008345
101443-782EA 149.08 USD
101443-782
Disodium malonate 2 M
Disodium malonate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: C₃H₂Na₂O₄
MW: 148.03 g/mol
MDL Number: MFCD00002708
CAS Number: 141-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR