Magnesium acetate 2 M

Supplier: Rigaku Reagents

1008335
101443-722EA 149.08 USD
101443-722
Magnesium acetate 2 M
Magnesium acetate solutions

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: (H₃CCOO)₂Mg·4H₂O
MDL Number: MFCD00149214
CAS Number: 16674-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR