1,6-Hexanediol 50% (w/v)

Supplier: Rigaku Reagents

1008029
101443-644EA 98.08 USD
101443-644
1,6-Hexanediol 50% (w/v)
1,6-Hexanediol

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: C₆H₁₄O₂
MDL Number: MFCD00002985
CAS Number: 629-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR