2-Ethoxyethanol ≥99% (v/v)

Supplier: Rigaku Reagents

1008400 1008401
101443-280EA 66.69 USD
101443-280 101443-614
2-Ethoxyethanol ≥99% (v/v)
2-Ethoxyethanol

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: C₂H₅OCH₂CH₂OH
MDL Number: MFCD00002869
CAS Number: 110-80-5
UN: 1171
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=461a4daf8709ebd6b4516d73b4b32eee33cd1bca&vid=625adefb8d1e2126363931c57a51d3e1df419926&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR