tri-Sodium citrate solution 1.6 M

Supplier: Rigaku Reagents

1008082 1008083
101443-386EA 82.38 USD
101443-386 101443-744
tri-Sodium citrate solution 1.6 M
tri-Sodium citrate solutions

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: Na₃C₆H₅O₇·2H₂O
MDL Number: MFCD00150031
CAS Number: 6132-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR