Lithium sulphate 2 M

Supplier: Rigaku Reagents

1008332
101443-352EA 105.92 USD
101443-352
Lithium sulphate 2 M
Lithium sulphate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.


Formula: Li₂SO₄
MDL Number: MFCD00011086
CAS Number: 10377-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR