Potassium sulfate 0.5 M

Supplier: Rigaku Reagents

1008043 1008042
101443-686EA 125.54 USD
101443-686 101443-338
Potassium sulfate 0.5 M
Potassium sulphate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
MDL Number: MFCD00011388
CAS Number: 7778-80-5
Merck Index: 13,07759

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR