JEFFAMINE® M-600 amine 50% (v/v) pH 7.0

Supplier: Rigaku Reagents

1008036
101443-312EA 117.69 USD
101443-312
JEFFAMINE® M-600 amine 50% (v/v) pH 7.0
JEFFAMINE® M-600 amine
Formula: CH₃CH(NH₂)CH₂[OCH(CH₃)CH₂]l(OCH₂CH₂)m[OCH₂CH(CH₃)]nNH₂
MDL Number: MFCD00147774
CAS Number: 77110-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR