Diammonium-L(+)-tartrate 1 M

Supplier: Rigaku Reagents

1008099 1008100
101443-274EA 79.77 USD
101443-274 101443-608
Diammonium-L(+)-tartrate 1 M
di-Ammonium L(+)-tartrate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: C₄H₁₂N₂O₆
MDL Number: MFCD00013073
CAS Number: 3164-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR