Cadmium(II) chloride 1 M

Supplier: Rigaku Reagents

1008153
101443-260EA 187 USD
101443-260
Cadmium(II) chloride 1 M
Cadmium (II) chloride

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: CdCl₂
MDL Number: MFCD00010916
CAS Number: 10108-64-2
UN: 2570
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR