Triammonium citrate 1 M

Supplier: Rigaku Reagents

Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate

1008111
101443-216EA 66.69 USD
101443-216
Triammonium citrate 1 M
tri-Ammonium citrate

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: C₆H₁₇N₃O₇
MW: 243.22 g/mol
MDL Number: MFCD00036406
CAS Number: 3458-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR