Tricyclopentylphosphine 10% (w/w) in hexanes ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

15-6181-50G
101442-480EA 319.88 USD
101442-480
Tricyclopentylphosphine 10% (w/w) in hexanes ≥95%
Tricyclopentylphosphine

Phosphine

MDL Number: MFCD00269834
CAS Number: 7650-88-6
UN: 1993
ADR: 3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR