Europium(III) trifluoromethanesulfonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Europium(III) trifluoromethanesulphonate, Europium triflate, Europium(III) trifluoromethanesulfonate

63-1000-1G 63-1000-5G
101442-370EA 46.71 USD
101442-370 101442-372
Europium(III) trifluoromethanesulfonate ≥98%
Europium (III) trifluoromethanesulphonate

Metal Triflate

Formula: C₃EuF₉O₉S₃
MDL Number: MFCD00209587
CAS Number: 52093-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR