Gold, nanoparticles (sugar coated) [AuNP: 6-10 nm (Maltose)]

Supplier: STREM CHEMICALS INC MS
Warning

79-0122-5ML
101442-308EA 942.65 USD
101442-308
Gold, nanoparticles (sugar coated) [AuNP: 6-10 nm (Maltose)]
Gold

Nanomaterials

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR