Gold, nanoparticles (water soluble) [AuNP: 3nm (Citrate)]

Supplier: STREM CHEMICALS INC MS
Warning

79-0110-100ML 79-0110-25ML
101442-288EA 867.63 USD
101442-288 101442-290
Gold, nanoparticles (water soluble) [AuNP: 3nm (Citrate)]
Gold

Nanomaterials

Formula: Au
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR