Gold, nanoparticles (water soluble) [AuNP: 10 nm (Citrate)]
Supplier: STREM CHEMICALS INC MS
|
Warning
|
79-0114-25ML
79-0114-100ML
101442-270EA
278.83
USD
101442-270
101442-268
Gold, nanoparticles (water soluble) [AuNP: 10 nm (Citrate)]
Gold
Nanomaterials
Formula:
Au |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...