1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,2-Bis(phenylsulphinyl)ethanepalladium(II) acetate, 1,2-Bis(phenylsulfinyl)ethanepalladium(II) acetate, 1,2-Bis(phenylsulfinyl)ethane palladium(II) acetate, 1,2-Bis(phenylsulphinyl)ethane palladium(II) acetate, White Catalyst, Christina White Catalyst

46-0245-1G 46-0245-250MG
101442-210EA 256.18 USD
101442-210 101442-208
1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate ≥98%
1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate
Formula: C₁₆H₁₈O₄PdS₂
MDL Number: MFCD09842752
CAS Number: 858971-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR