1,2-Bis(di-2-pyridylphosphino)ethane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-0683-100MG 15-0683-500MG
101442-204EA 131.63 USD
101442-204 101442-206
1,2-Bis(di-2-pyridylphosphino)ethane ≥98%
1,2-Bis(di-2-pyridylphosphino)ethane

Phosphine

MDL Number: MFCD10565625
CAS Number: 106308-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR