(1R,2R)-[1,2-Cyclohexanediamino-N,N'-bis(4-bromobenzyl)allylchlorosilane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

14-1880-250MG 14-1880-1G
101442-162EA 137.29 USD
101442-162 101442-164
(1R,2R)-[1,2-Cyclohexanediamino-N,N'-bis(4-bromobenzyl)allylchlorosilane ≥98%
(1R,2R)-[1,2-Cyclohexanediamino-N,N'-bis(4-bromobenzyl)allylchlorosilane

Organocatalyst

MDL Number: MFCD10567380
CAS Number: 546084-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR