Lab Aid® Ultra™ II Storage Cabinet, Sakura
Supplier: Sakura Finetek
3990
10144-724EA 7985.63 USD
10144-724
Lab Aid® Ultra™ II Storage Cabinet, Sakura
Furniture Storage Cabinets
Security and capacity are the primary characteristics of the Lab Aid® Ultra™ cabinet for slides or cassettes.
  • Storage capacity is either 96,000 slides or 22,000 cassettes (blocks)
  • Two keys to lock the drawers and hardware to connect cabinets together
10 drawer cabinet; eight trays with eight dividers in each drawer for a total of 80 trays and dividers per cabinet.
This modernized system offers large capacity storage with integrated security features. A sturdy steel construction ensures long-term use and a drawer lock prevents tampering, guaranteeing specimen safety. Each drawer front has a plastic covered labeling tab for positive identification.