Tissue Cassette Storage Cabinet, Boekel
Supplier: Boekel
143000
10144-720EA
212.5
USD
10144-720
Tissue Cassette Storage Cabinet, Boekel
Furniture
Storage Cabinets
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...