Micro-organism Lysing VKMix, Precellys® Lysing Kit, Bertin
Supplier: BERTIN CORP
P000937-LYSK0-A.0
10144-530EA 368.41 USD
10144-530
Micro-organism Lysing VKMix, Precellys® Lysing Kit, Bertin
Beads Glass Beads
VKMix is made with inert 0.1 mm and 0.5 mm glass beads. It is designed for micro-organism lysing like yeast fungi, gram+, gram- bacteria, spores, micro-algae and more.
  • Ideal for yeast, bacteria and fungi lysis
  • Dnase/Rnase free
  • Autoclavable
Use with the Precellys® Evolution, or Minilys homogenizers.
50 tubes (prefilled)
Certifications: Dnase/Rnase free certified.
Ordering information: Only compatible with Precellys® Evolution and Minilys® homogenizers.
Packaging: Packaged 1 kit contain 50 prefilled and ready-to-use tubes.