6-Outlet Hospital-Grade Surge Protector, Tripp Lite
Supplier: Tripp Lite
SPS606HGRA
10144-446EA 428.19 USD
10144-446
6-Outlet Hospital-Grade Surge Protector, Tripp Lite
Surge Suppressors
This surge suppressor provides six hospital grade outlets and protection from damaging surges and spikes with the added benefit of a convenient right-angle hospital grade plug.
  • AC surge suppression rating of 1050 joules
  • Rugged all-metal housing withstands years of use without loss of ground path continuity
  • Six NEMA5-15R hospital-grade AC outlets
  • Key hole mounting tabs and flanges
  • Switch safety cover prevents accidental shutoff
  • 15 A electrical capacity with built-in resettable circuit breaker
The new design provides another option for convenient outlets outside the patient care area, including those tight spaces.
Not for patient-care rooms.
Accessories information: HG outlet cover compatible with all NEMA 5-15R-HG and NEMA 5-15R outlets.
Caution: NOT for patient-care rooms. Not for use where NEC requires hospital-grade receptacles.