LiChroCART® Purospher® HPLC Columns, STAR RP-8e, MilliporeSigma
Supplier: MilliporeSigma
1.50171.7194
10144-056EA 727.39 USD
10144-056
LiChroCART® Purospher® HPLC Columns, STAR RP-8e, MilliporeSigma
Chromatography Columns
Stainless steel LiChroCART® cartridges are available with various types of Purospher® sorbents.
These LiChroCART® cartridges require part number 14229-736 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a 4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 48219-720 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of 3 cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 48219-758 LiChroCART® cartridge holder for 30 mm cartridge and 48219-756 LiChroCART® cartridge holder for 55 mm cartridge.