Gold, nanoparticles (water soluble) [AuNP: 5nm (Citrate)]

Supplier: STREM CHEMICALS INC MS
Warning

79-0112-100ML 79-0112-25ML
101433-260EA 842.15 USD
101433-260 101433-262
Gold, nanoparticles (water soluble) [AuNP: 5nm (Citrate)]
Gold

Nanomaterials

Formula: Au
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR