Cerium(IV) oxide, nanopowder

Supplier: STREM CHEMICALS INC MS

Synonyms: Ceric oxide, Cerium dioxide

58-1400-100G 58-1400-25G
101433-256EA 274.58 USD
101433-256 101433-258
Cerium(IV) oxide, nanopowder
Cerium (IV) oxide

Nanomaterials

Formula: CeO₂
Boiling Pt: 3468 °C (1013 hPa)
Melting Pt: 2000 °C
Density: 7.13 g/cm³ (25 °C)
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR