(9-Benzyl-9-fluorenyl)dicyclohexylphosphonium tetrafluoroborate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: cataCXium® FBn, Dicyclohexyl(9-benzylfluoren-9-yl)phosphonium tetrafluoroborate

15-1072-500MG 15-1072-2G
101433-122EA 157.11 USD
101433-122 101433-124
(9-Benzyl-9-fluorenyl)dicyclohexylphosphonium tetrafluoroborate ≥97%
(9-Benzyl-9-fluorenyl)dicyclohexylphosphonium tetrafluoroborate

cataCXium

Formula: C₃₂H₃₈BF₄P
MDL Number: MFCD10567010
CAS Number: 937378-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR