6,6'-[(3,3'-Di-t-butyl-5,5'-dimethoxy-1,1'-biphenyl-2,2'-diyl)bis(oxy)] bis(dibenzo[d,f][1,3,2]dioxaphosphepin) hemi ethyl acetate adduct ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

15-0107-1G 15-0107-250MG
101433-100EA 496.8 USD
101433-100 101433-098
6,6'-[(3,3'-Di-t-butyl-5,5'-dimethoxy-1,1'-biphenyl-2,2'-diyl)bis(oxy)] bis(dibenzo[d,f][1,3,2]dioxaphosphepin) hemi ethyl acetate adduct ≥95%
6,6'-[(3,3'-Di-t-butyl-5,5'-dimethoxy-1,1'-biphenyl-2,2'-diyl)bis(oxy)] bis(dibenzo[d,f][1,3,2]dioxaphosphepin) hemi ethyl acetate adduct

Phosphine

Formula: C₄₆H₄₄O₈P₂
Melting Pt: 188…190 °C
MDL Number: MFCD09839135
CAS Number: 121627-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR