RediSep Rf Silica Flash Chromatography Column, Teledyne ISCO
Supplier: Teledyne
692203304 692203275 692203312 692203380 692203320 692203330 692203277 692203324 692203340 692203422 692203527
101432-862PK 107.69 USD
101432-862 101432-858 101432-864 101432-922 101432-866 101432-870 101432-860 101432-868 101432-872 101432-972 101671-818
RediSep Rf Silica Flash Chromatography Column, Teledyne ISCO
Chromatography Columns
RediSep Rf disposable, bare silica columns for flash purification of organic compounds are the benchmark for normal phase separations.
This high quality media is specified for high resolution and reproducibility for everyday purifications. Reusability: Single use.