Papanicolaou's solution EA 36, VWR® stain for histology, for cytology
Supplier: VWR International
|
Danger
|
7042-32
10143-616CS
3150.32
USD
10143-616
Papanicolaou's solution EA 36, VWR® stain for histology, for cytology
Papanicolaou's solution EA 36
Papanicolaou stains are used in conjunction with Hematoxylin nuclear stains in the diagnosis of malignant cytological diseases.
- Crisp, clear staining
- Well-defined cytoplasm
- ready-to-use
Gynecological staining protocols use OG-6 and hematoxylin along with either EA-36 or EA-50. Non gynecological procedures require use of EA-65. EASYpap is a single step cytology staining solution. Combination of EA and OG-6 solutions saves times by providing customer with an easy one step process. FDA approved for in vitro diagnostic use. For Professional Use Only.
Storage Temperature:
Ambient |
UN:
1170 ADR: 3,II |
Specification Test Results
Absorbance (Visible Scan 700- 400nm) @626 nm | 270 – 350 MA |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...