Wright-Giemsa stain, VWR® Quick III™ for hematology

Supplier: VWR International

Synonyms: Wright-Giemsa Solution

10143-236
10143-236CS 1739.36 USD
10143-236
Wright-Giemsa stain, VWR® Quick III™ for hematology
Wright-Giemsa stain (solutions & kits)

VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.


  • Stain offerings for multiple protocols
  • Individual stains, reagents and packs have been used in the automated market on the Hematek®, Midas®, Sysmex® and Sakura® instruments


Single-step stains are offered in Wright's and Wright Giemsa formulations and use deionized water to complete the staining process. Three step stain offering yields results comparable to Wright Giemsa stain with results available as soon as 15 seconds.


Certifications: Hematology stain products are manufactured using dyes certified by the Biological Stain Commission. All products undergo Quality Control evaluation using fresh blood.

Order Now

Specification Test Results

Quick III Solution I Absorbance 300-340MA @~518nm
Quick III Solution I pH 6.55-6.65@25°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR