Wright-Giemsa stain, VWR® Quick III™ Set for hematology
Supplier: VWR International
Synonyms:
Wright-Giemsa Solution
VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.
- Stain offerings for multiple protocols
- Individual stains, reagents and packs have been used in the automated market on the Hematek®, Midas®, Sysmex® and Sakura® instruments
Single-step stains are offered in Wright's and Wright Giemsa formulations and use deionized water to complete the staining process. Three step stain offering yields results comparable to Wright Giemsa stain with results available as soon as 15 seconds.
Certifications: Hematology stain products are manufactured using dyes certified by the Biological Stain Commission. All products undergo Quality Control evaluation using fresh blood.
Specification Test Results
Quick III Solution I Absorbance | 300-340MA @~518nm |
Quick III Solution I pH | 6.55-6.65@25°C |
Quick III Solution II Absorbance | 440-460MN@~650nm |
Quick III Solution II pH | 6.85-6.95@25°C |
Quick III Fixative | Physical Inspection |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...