Wright's stain, VWR® for hematology

Supplier: VWR International

Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue

5580-32 5580-G
10143-164CS 2697.94 USD
10143-164 10143-166
Wright's stain, VWR® for hematology
Wright's stain

VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.


  • Stain offerings for multiple protocols
  • Individual stains, reagents and packs have been used in the automated market on the Hematek®, Midas®, Sysmex® and Sakura® instruments


Single-step stains are offered in Wright's and Wright Giemsa formulations and use deionized water to complete the staining process. Three step stain offering yields results comparable to Wright Giemsa stain with results available as soon as 15 seconds.


Certifications: Hematology stain products are manufactured using dyes certified by the Biological Stain Commission. All products undergo Quality Control evaluation using fresh blood.

Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1

Order Now

Specification Test Results

Absorbance Eosin Visible Scan-(700- 400nm) 325– 375 MA@525 nm
Absorbance MB/Azure Visible Scan (700-400nm) 580-680 MA @650 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR