Hematoxylin solution Gill I, VWR® stain for histology, for cytology
Supplier: VWR International
|
Warning
|
Gill 1X is ideal for routine cytology staining. This single strength formulation optimally stains gynecological and non-gynecological specimens.
- Crisp, clear staining
- Well-defined cytoplasm
- ready-to-use
General purpose nuclear stain, progressive type. Used with hematoxylin and eosin staining. Gill 2X and Gill 3X should be used when a stronger or darker nuclear stain is required for histology samples. FDA approved for in vitro diagnostic use. For Professional Use Only.
Formula:
C₁₆H₁₄O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Specification Test Results
Absorbance (Visible Scan 700- 400nm) @~560 nm | ≥125 MA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...