Wright's stain PK, VWR® for hematology
Supplier: VWR International
Synonyms:
Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.
- Stain offerings for multiple protocols
- Individual stains, reagents and packs have been used in the automated market on the Hematek®, Midas®, Sysmex® and Sakura® instruments
Single-step stains are offered in Wright's and Wright Giemsa formulations and use deionized water to complete the staining process. Three step stain offering yields results comparable to Wright Giemsa stain with results available as soon as 15 seconds.
Certifications: Hematology stain products are manufactured using dyes certified by the Biological Stain Commission. All products undergo Quality Control evaluation using fresh blood.
Storage Temperature:
Ambient |
MDL Number:
MFCD00082143 CAS Number: 68988-92-1 |
Specification Test Results
Wright’s absorbance | Azure 0.580-0.680@650nm |
Buffer pH | 6.65-6.75 |
Rinse pH | 6.45-6.55 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...