Sodium chloride USP

Supplier: HAWKINS PC
Warning

Synonyms: Saline, NaCl, Sea salt, Halite

14703
10142-840EA 249.04 USD
10142-840
Sodium chloride USP
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR