Pierceable Plug Stopper - For 13 mm Tubes, Globe Scientific
Supplier: Globe Scientific
113150B 113150G 113150R 113150W 113150Y
10142-466PK 274.84 USD
10142-466 10142-468 10142-470 10142-472 10142-474
Pierceable Plug Stopper - For 13 mm Tubes, Globe Scientific
Stoppers Rubber Stoppers
These pierceable plug stoppers are intended for recapping vacuum blood tubes and test tubes.
  • Thermoplastic elastomer (TPE)
  • Assorted colors available
  • Fits both glass and plastic tubes
The caps are produced from thermoplastic elastomer (TPE) which can easily be pierced with a syringe.
Ordering information: Stoppers are packaged 1000 per bag.