Takasago BINAP Ru Dimer Catalyst Kit
Supplier: STREM CHEMICALS INC MS
96-6952-1KIT
101419-194EA 457.17 USD
101419-194
Takasago BINAP Ru Dimer Catalyst Kit
Takasago BINAP Ru Dimer Catalyst Kit
BINAP
This Kit Contains 250mg each of the following Strem Product #s: 44-0510, 44-0511, 44-0512, 44-0513, 44-0514, 44-0515.
Please check strem.com for any updates and changes.