2,5-Norbornadienepalladium(II) dichloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (Bicyclo[2.2.1]hepta-2,5-diene)dichloropalladium(ii), Dichloro(norbornadiene)palladium(II), 2,5-Norbornadienedichloropalladium(II), Dichloro-2,5-norbornadiene Palladium(II)

46-0900-250MG 46-0900-1G
101419-072EA 60.86 USD
101419-072 101419-074
2,5-Norbornadienepalladium(II) dichloride ≥99%
2,5-Norbornadienepalladium(II) dichloride
Formula: C₇H₈Cl₂Pd
MDL Number: MFCD00134222
CAS Number: 12317-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR