(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

45-2110-100MG
101419-054EA 181.17 USD
101419-054
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥97%
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

P-PHOS

MDL Number: MFCD09264281
CAS Number: 573718-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR