RuCl2[(R)-xylbinap][(R,R)-dpen]

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dichloro[(R)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl][(R,R)-1,2-diphenylethylenediamine]ruthenium(II), Dichloro{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II) RuCl2[(R)-xylbinap][(R,R)-dpen]

44-0226-250MG
101418-950EA 90.58 USD
101418-950
RuCl2[(R)-xylbinap][(R,R)-dpen]
RuCl2[(R)-xylbinap][(R,R)-dpen]

BINAP

Formula: C₆₆H₆₄Cl₂N₂P₂Ru
MDL Number: MFCD09753028
CAS Number: 220114-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR