Diacetato{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}ruthenium(II)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ru(OAc)2[(S)-dm-segphos®]

44-0176-250MG
101418-922EA 90.58 USD
101418-922
Diacetato{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}ruthenium(II)
Diacetato{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}ruthenium(II)

SEGPHOS

Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753025
CAS Number: 944450-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR