Diacetato{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}ruthenium(II)

Supplier: STREM CHEMICALS INC MS
Warning

44-0164-250MG
101418-890EA 90.58 USD
101418-890
Diacetato{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}ruthenium(II)
Diacetato{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}ruthenium(II)

BINAP

Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753022
CAS Number: 374067-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR