Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ru(OAc)2[(R)-binap]

44-0152-250MG 44-0152-1G
101418-872EA 75.02 USD
101418-872 101418-874
Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)

BINAP

Formula: C₄₈H₃₈O₄P₂Ru
Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753020
CAS Number: 325146-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR