(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-SIPHOS, (S)-N,N-Dimethyl-1,2,4,5,6,7-hexahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine

15-5151-100MG 15-5151-500MG
101418-672EA 157.11 USD
101418-672 101418-674
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine ≥98%
(11aS)-(-)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-dimethylamine

MONOPHOS, Phosphine, SIPHOS

Formula: C₁₉H₂₀NO₂P
Melting Pt: 95…96 °C
MDL Number: MFCD08459340
CAS Number: 443965-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR