(S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0489-100MG 15-0489-500MG
101418-590EA 140.12 USD
101418-590 101418-592
(S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥97%
(S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl

BIPHEP, Phosphine

Formula: C₄₆H₄₈O₂P₂
MDL Number: MFCD09753009
CAS Number: 362634-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR