(S)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-(-)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole, (S)-(-)-DM-SEGPHOS®, (S)-(-)-4,4'-Bis[di(3,5-xylyl)phosphino]-3,3'-bi(1,2-methylenedioxybenzene)

15-0479-250MG
101418-584EA 67.94 USD
101418-584
(S)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole ≥98%
(S)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole

Phosphine, SEGPHOS

Formula: C₄₆H₄₄O₄P₂
Melting Pt: 256…261 °C
MDL Number: MFCD09753008
CAS Number: 210169-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR