Low Range ssRNA Ladder, New England Biolabs
Supplier: New England Biolabs (NEB)
N0364S
101418-104EA 84 USD
101418-104
Low Range ssRNA Ladder, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
The Low Range ssRNA Ladder is a set of 6 RNA molecules produced by in vitro transcription of a mixture of 6 linear DNA templates
The ladder sizes are: 1000, 500, 300, 150, 80 and 50 bases. The 300 base fragment is at double intensity to serve as a reference band. This ladder is suitable for use as an ssRNA size standard on denaturing polyacrylamide-urea gels, and on denaturing or native agarose gels.